missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. D05651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05651G 1 g
D05655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05651G Supplier TCI America Supplier No. D05651G

Chemical Identifiers

CAS 1194-98-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003333
InChI Key CLFRCXCBWIQVRN-UHFFFAOYSA-N
Synonym gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
PubChem CID 70949
ChEBI CHEBI:28508
IUPAC Name 2,5-dihydroxybenzaldehyde
SMILES OC1=CC=C(O)C(C=O)=C1

Specifications

Melting Point 102°C
Quantity 1 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxybenzaldehyde