missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzaldehyde, 99%

Catalog No. AC114840050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114840050 5 g Glass bottle
AC114840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC114840050 Supplier Thermo Scientific Chemicals Supplier No. 114840050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1194-98-5 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1194-98-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003333
InChI Key CLFRCXCBWIQVRN-UHFFFAOYSA-N
Synonym gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy
PubChem CID 70949
ChEBI CHEBI:28508
IUPAC Name 2,5-dihydroxybenzaldehyde
SMILES OC1=CC=C(O)C(C=O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color Green-Khaki to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 5 g
Beilstein 08,244
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 138.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dihydroxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 214-789-6

RUO – Research Use Only