missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydroxy-1,4-dithiane 96.0+%, TCI America™
Supplier: TCI America D1761500G
Specifications
| 2,5-Dihydroxy-1,4-dithiane | |
| White-Yellow | |
| MFCD00006659 | |
| p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme | |
| C1C(SCC(S1)O)O | |
| 152.226 | |
| 152.23 | |
| Crystalline Powder |
| 40018-26-6 | |
| C4H8O2S2 | |
| 500 g | |
| YUIOPHXTILULQC-UHFFFAOYSA-N | |
| 1,4-dithiane-2,5-diol | |
| 98330 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 40018-26-6 | |
| 152.226 | |
| YUIOPHXTILULQC-UHFFFAOYSA-N | |
| 98330 | |
| C1C(SCC(S1)O)O |
| C4H8O2S2 | |
| MFCD00006659 | |
| p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme | |
| 1,4-dithiane-2,5-diol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator