missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-1,4-dithiane 96.0+%, TCI America™

Catalog No. D176125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176125G 25 g
D1761500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D176125G Supplier TCI America Supplier No. D176125G

Chemical Identifiers

CAS 40018-26-6
Molecular Formula C4H8O2S2
Molecular Weight (g/mol) 152.226
MDL Number MFCD00006659
InChI Key YUIOPHXTILULQC-UHFFFAOYSA-N
Synonym p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
PubChem CID 98330
IUPAC Name 1,4-dithiane-2,5-diol
SMILES C1C(SCC(S1)O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 152.23
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxy-1,4-dithiane
TSCA No
Recommended Storage Refrigerator