missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-1,4-benzoquinone, 98%

Catalog No. p-7081457
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1144206 5 g
AAA1144214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1144206 Supplier Thermo Scientific Chemicals Supplier No. A1144206
Only null left

CAS: 615-94-1 | C6H4O4 | 140.09 g/mol

2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 615-94-1
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00001598
InChI Key QFSYADJLNBHAKO-UHFFFAOYSA-N
Synonym 2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop
PubChem CID 69213
IUPAC Name 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC(=O)C(O)=CC1=O

Specifications

Melting Point ∼220°C (sublimation)
Quantity 5 g
Beilstein 1939229
Solubility Information Soluble in water.
Formula Weight 140.1
Percent Purity 98%
Chemical Name or Material 2,5-Dihydroxy-1,4-benzoquinone
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-454-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only