accessibility menu, dialog, popup

UserName

OHOOOH

CAS RN 615-94-1

IUPAC Name: 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
Synonyms: 2,5-dihydroxy-1,4-benzoquinone
Molecular Weight (g/mol): 140.09
Molecular Formula: C6H4O4
InChi Key: QFSYADJLNBHAKO-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C(O)=CC1=O
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)