missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic Acid 97.0+%, TCI America™

Catalog No. D39555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39555G 5 g
D39551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39555G Supplier TCI America Supplier No. D39555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 487-66-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.15
MDL Number MFCD18252872
InChI Key FWBJLYXRIRBVQG-UHFFFAOYSA-N
Synonym 2-(2-Carboxyethyl)-3-methylmaleic Anhydride
PubChem CID 11183025
IUPAC Name 3-(4-methyl-2,5-dioxo-2,5-dihydrofuran-3-yl)propanoic acid
SMILES CC1=C(CCC(O)=O)C(=O)OC1=O

Specifications

Melting Point 99°C
Color White
Quantity 5 g
Formula Weight 184.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic Acid