accessibility menu, dialog, popup

UserName

CAS RN 487-66-1

IUPAC Name: 3-(4-methyl-2,5-dioxo-2,5-dihydrofuran-3-yl)propanoic acid
Synonyms: 3-(4-methyl-2,5-dioxofuran-3-yl)propanoic acid
Molecular Weight (g/mol): 184.15
Molecular Formula: C8H8O5
InChi Key: FWBJLYXRIRBVQG-UHFFFAOYSA-N
SMILES: CC1=C(CCC(O)=O)C(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results