missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Catalog No. p-7027950
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2502822 100 g
AAB2502830 250 g
AAB2502836 500 g
AAB250280B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAB2502822 Supplier Thermo Scientific Chemicals Supplier No. B2502822
Only null left

CAS: 332-77-4 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 332-77-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00003220
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
Synonym 2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
PubChem CID 78974
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
SMILES COC1OC(OC)C=C1

Specifications

Melting Point <-40°C
Density 1.072
Boiling Point 160°C to 162°C
Flash Point 47°C (116°F)
Refractive Index 1.435
Quantity 100 g
UN Number UN3271
Beilstein 113225
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans
Show More Show Less
missing translation for 'healthHazard1' H226-H302+H332-H319
missing translation for 'healthHazard2' GHS H Statement
H301-H226-H332
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.
missing translation for 'healthHazard3' P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 206-367-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only