missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Diformylfuran 98.0+%, TCI America™
Supplier: TCI America D24085G
Specifications
| 2,5-Diformylfuran | |
| 110°C | |
| MFCD00671517 | |
| 2,5-Furandicarboxaldehyde | |
| O=CC1=CC=C(O1)C=O | |
| 124.10 | |
| CHEBI:83385 | |
| ≥98.0% (GC) |
| 823-82-5 | |
| C6H4O3 | |
| 5 g | |
| PXJJKVNIMAZHCB-UHFFFAOYSA-N | |
| furan-2,5-dicarbaldehyde | |
| 69980 | |
| 124.10 | |
| Crystalline Powder |
Chemical Identifiers
| 823-82-5 | |
| 124.10 | |
| PXJJKVNIMAZHCB-UHFFFAOYSA-N | |
| 69980 | |
| furan-2,5-dicarbaldehyde |
| C6H4O3 | |
| MFCD00671517 | |
| 2,5-Furandicarboxaldehyde | |
| CHEBI:83385 | |
| O=CC1=CC=C(O1)C=O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator