missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diformylfuran 98.0+%, TCI America™

Catalog No. p-7127282
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24081G 1 g
D24085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24081G Supplier TCI America Supplier No. D24081G
Only null left

Chemical Identifiers

CAS 823-82-5
Molecular Formula C6H4O3
Molecular Weight (g/mol) 124.10
MDL Number MFCD00671517
InChI Key PXJJKVNIMAZHCB-UHFFFAOYSA-N
Synonym 2,5-Furandicarboxaldehyde
PubChem CID 69980
ChEBI CHEBI:83385
IUPAC Name furan-2,5-dicarbaldehyde
SMILES O=CC1=CC=C(O1)C=O

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 124.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diformylfuran
TSCA No
Recommended Storage Refrigerator