missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7127731
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34365G 5 g
D343625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34365G Supplier TCI America Supplier No. D34365G
Only null left

Chemical Identifiers

CAS 193353-34-3
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD01863171
InChI Key KTOJGSDLJNUAEP-UHFFFAOYSA-N
Synonym 2,5-difluorophenyl boronic acid, 2,5-difluorobenzeneboronic acid, 2,5-difluorophenyl boranediol, boronic acid, 2,5-difluorophenyl, 2,5-difluorophenylboronicacid, 2,5-fluorobenzene boronic acid, pubchem1820, acmc-1c1am, ksc174i6r, 2,5-difluoro phenylboronic acid
PubChem CID 2734335
IUPAC Name (2,5-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)F)F)(O)O

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Formula Weight 157.91
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Difluorophenylboronic Acid (contains varying amounts of Anhydride)