missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenol 95.0+%, TCI America™

Catalog No. D20365G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20365G Supplier TCI America Supplier No. D20365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2713-31-7
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00042501
InChI Key INXKVYFOWNAVMU-UHFFFAOYSA-N
Synonym phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464
PubChem CID 94952
IUPAC Name 2,5-difluorophenol
SMILES OC1=CC(F)=CC=C1F

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 145°C
Quantity 5 g
UN Number 1325
Formula Weight 130.09
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,5-Difluorophenol