Learn More
2,5-Difluorophenol, 97%
CAS: 2713-31-7 | C6H4F2O | 130.09 g/mol
$222.42 - $222.42
Chemical Identifiers
| CAS | 2713-31-7 |
|---|---|
| Molecular Formula | C6H4F2O |
| Molecular Weight (g/mol) | 130.09 |
| MDL Number | MFCD00042501 |
| InChI Key | INXKVYFOWNAVMU-UHFFFAOYSA-N |
| Synonym | phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464 |
| PubChem CID | 94952 |
| IUPAC Name | 2,5-difluorophenol |
| SMILES | OC1=CC(F)=CC=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1874414
|
Thermo Scientific Chemicals
A1874414 |
25 g |
Each for $222.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2713-31-7 | |
| 130.09 | |
| INXKVYFOWNAVMU-UHFFFAOYSA-N | |
| 94952 | |
| OC1=CC(F)=CC=C1F |
| C6H4F2O | |
| MFCD00042501 | |
| phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464 | |
| 2,5-difluorophenol |
Specifications
| 2713-31-7 | |
| 53°C (127°F) | |
| C6H4F2O | |
| 25 g | |
| 2081076 | |
| INXKVYFOWNAVMU-UHFFFAOYSA-N | |
| 2,5-difluorophenol | |
| 94952 | |
| 97% |
| 36°C to 40°C | |
| Phenol-like | |
| MFCD00042501 | |
| UN2921 | |
| phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464 | |
| OC1=CC(F)=CC=C1F | |
| 130.09 | |
| 130.09 | |
| 2,5-Difluorophenol |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302+H312+H332-H314
DOTInformation : Hazard Class: 4.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only