missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D34065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34065G 5 g
D340625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34065G Supplier TCI America Supplier No. D34065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35730-09-7
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00009929
InChI Key RLRUKKDFNWXXRT-UHFFFAOYSA-N
PubChem CID 588082
IUPAC Name 2,5-difluorobenzoyl chloride
SMILES C1=CC(=C(C=C1F)C(=O)Cl)F

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 2920
Synonym 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
Formula Weight 176.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzoyl Chloride