missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoyl chloride, 97+%

Catalog No. AAA1065114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065114 25 g
AAA1065106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1065114 Supplier Thermo Scientific Chemicals Supplier No. A1065114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35730-09-7 | C7H3ClF2O | 176.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35730-09-7
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00009929
InChI Key RLRUKKDFNWXXRT-UHFFFAOYSA-N
Synonym 2,5-difluorobenzoylchloride, benzoyl chloride, 2,5-difluoro, benzoyl chloride,2,5-difluoro, acmc-1afez, 2,5-difluoro benzoyl chloride, 2,5-difluoro-benzoyl chloride, 2,5-difluorobenzoyl chloride
PubChem CID 588082
IUPAC Name 2,5-difluorobenzoyl chloride
SMILES C1=CC(=C(C=C1F)C(=O)Cl)F

Specifications

Density 1.425
Boiling Point 81°C to 83°C (14 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.515
Quantity 25 g
UN Number UN3265
Beilstein 2046666
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzoyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-701-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only