missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzaldehyde 96.0+%, TCI America™

Catalog No. D26375G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26375G 5 g
D263725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. D26375G missing translation for 'mfr' TCI America missing translation for 'supplierNo' D26375G

Chemical Identifiers

CAS 2646-90-4
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010327
InChI Key VVVOJODFBWBNBI-UHFFFAOYSA-N
Synonym 2,5-difluoro benzaldehyde, 2,5-difluoro-benzaldehyde, 2,5-difluorobenzaldehyde, benzaldehyde, 2,5-difluoro, timtec-bb sbb006570, 2,5-difluorobenzalde, pubchem1437, 2,5-fluorobenzaldehyde, 2,5 difluorobenzaldehyde, acmc-1cq4a
PubChem CID 137663
IUPAC Name 2,5-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)F

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
UN Number 1989
Formula Weight 142.10
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzaldehyde