missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-nitrobenzoic acid, 97%

Catalog No. p-7052449
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3375703 1 g
AAH3375706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3375703 Supplier Thermo Scientific Chemicals Supplier No. H3375703
Only null left

CAS: 116465-48-6 | C7H3F2NO4 | 203.101 g/mol

2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 116465-48-6
Molecular Formula C7H3F2NO4
Molecular Weight (g/mol) 203.101
MDL Number MFCD03425705
InChI Key GPTNSBLYGCZJQV-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-nitrobenzoic acid, 2,5-difluoro-4-nitrobenzenecarboxylic acid, 2,5-difluoro-4-nitrobenzic acid, 2,5-difluoro-4-nitrobenzoicacid, 2,5-difluoro-4-nitro-benzoic acid, benzoic acid, 2,5-difluoro-4-nitro, 2,5-difluoro-4-nitrobenzenecarboxylicacid, pubchem9789, acmc-2099sr, ksc495m6l
PubChem CID 246375
SMILES C1=C(C(=CC(=C1F)[N+](=O)[O-])F)C(=O)O

Specifications

Melting Point 145°C to 147°C
Color Green to White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 203.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only