missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-hydroxybenzaldehyde, 99%

CAS: 918523-99-6 | C7H4F2O2 | 158.104 g/mol

Supplier:  Thermo Scientific Chemicals H3152803

 View more versions of this product

Catalog No. AAH3152803


Only null left
Explore available promotions

Description

Description

2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
Specifications

Specifications

2,5-Difluoro-4-hydroxybenzaldehyde
145°C to 148°C
MFCD04112521
Air Sensitive
Slightly soluble in water.
C1=C(C(=CC(=C1F)O)F)C=O
158.104
158.11
918523-99-6
C7H4F2O2
1 g
4-hydroxy-2,5-difluorobenzaldehyde, 2,5-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,5-difluoro-4-hydroxy, ksc496a0b
FFPHRYLGIPAZBL-UHFFFAOYSA-N
2,5-difluoro-4-hydroxybenzaldehyde
21966462
99%

Chemical Identifiers

918523-99-6
158.104
FFPHRYLGIPAZBL-UHFFFAOYSA-N
21966462
C1=C(C(=CC(=C1F)O)F)C=O
C7H4F2O2
MFCD04112521
4-hydroxy-2,5-difluorobenzaldehyde, 2,5-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,5-difluoro-4-hydroxy, ksc496a0b
2,5-difluoro-4-hydroxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only