missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorotoluene, 98%

Catalog No. AAA1303914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1303914 25 g
AAA1303922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1303914 Supplier Thermo Scientific Chemicals Supplier No. A1303914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19398-61-9 | C7H6Cl2 | 161.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19398-61-9
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000609
InChI Key KFAKZJUYBOYVKA-UHFFFAOYSA-N
Synonym 2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
PubChem CID 29572
IUPAC Name 1,4-dichloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 4°C to 5°C
Density 1.256
Boiling Point 198°C to 200°C
Flash Point 79°C (174°F)
Refractive Index 1.547
Quantity 25 g
UN Number UN3082
Beilstein 1859112
Formula Weight 161.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorotoluene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 243-032-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only