accessibility menu, dialog, popup

UserName

CAS RN 19398-61-9

IUPAC Name: 1,4-dichloro-2-methylbenzene
Synonyms: 1,4-dichloro-2-methylbenzene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: KFAKZJUYBOYVKA-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)