missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophenol, 98%

Catalog No. p-7081430
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1143403 1 g
AAA1143406 5 g
AAA1143414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1143403 Supplier Thermo Scientific Chemicals Supplier No. A1143403

CAS: 5858-18-4 | C6H4Cl2S | 179.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5858-18-4
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00004831
InChI Key QIULLHZMZMGGFH-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophenol, 2,5-dichlorobenzene-1-thiol, benzenethiol, 2,5-dichloro, 2,5-dichloro thiophenol, thiophenol, 2,5-dichloro, 2,5-dichlorophenylmercaptan, pubchem6801, thiophenol,5-dichloro, 2,5-dichlorobenzenthiol, 2,5-dichloro-thiophenol
PubChem CID 79965
IUPAC Name 2,5-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)S)Cl

Specifications

Melting Point 22°C to 25°C
Boiling Point 112°C to 116°C (50 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.619
Quantity 1 g
Beilstein 2042866
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorothiophenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 227-486-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only