missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene 97.0+%, TCI America™

Catalog No. D025025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D025025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D025025G Supplier TCI America Supplier No. D025025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3172-52-9
Molecular Formula C4H2Cl2S
Molecular Weight (g/mol) 153.02
MDL Number MFCD00005423
InChI Key FGYBDASKYMSNCX-UHFFFAOYSA-N
Synonym thiophene, 2,5-dichloro, 2,-dichlorothiophene, 2,5-dichloro-thiophene, pubchem5521, thiophene,5-dichloro, 2,5-dichloro thiophene, thiophene,2,5-dichloro, acmc-1cmg3, ksc223o3n, bidd:gt0088
PubChem CID 18501
IUPAC Name 2,5-dichlorothiophene
SMILES C1=C(SC(=C1)Cl)Cl

Specifications

Boiling Point 162°C
Quantity 25 g
UN Number 2929
Formula Weight 153.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dichlorothiophene
TSCA Yes
Recommended Storage Refrigerator