CAS RN 3172-52-9
IUPAC Name:
2,5-dichlorothiophene
Synonyms:
2,5-dichlorothiophene
Molecular Weight (g/mol):
153.02
Molecular Formula:
C4H2Cl2S
InChi Key:
FGYBDASKYMSNCX-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(Cl)S1
1
–
1
of
1
results
1
2,5-Dichlorothiophene 97.0+%, TCI America™
CAS: 3172-52-9 Molecular Formula: C4H2Cl2S Molecular Weight (g/mol): 153.02 MDL Number: MFCD00005423 InChI Key: FGYBDASKYMSNCX-UHFFFAOYSA-N Synonym: thiophene, 2,5-dichloro,2,-dichlorothiophene,2,5-dichloro-thiophene,pubchem5521,thiophene,5-dichloro,2,5-dichloro thiophene,thiophene,2,5-dichloro,acmc-1cmg3,ksc223o3n,bidd:gt0088 PubChem CID: 18501 IUPAC Name: 2,5-dichlorothiophene SMILES: C1=C(SC(=C1)Cl)Cl