missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloroterephthalic Acid 97.0+%, TCI America™
$41.02 - $109.98
Chemical Identifiers
| CAS | 13799-90-1 |
|---|---|
| Molecular Formula | C8H4Cl2O4 |
| MDL Number | MFCD00058982 |
Chemical Identifiers
| 13799-90-1 | |
| C8H4Cl2O4 | |
| MFCD00058982 |
Specifications
| 13799-90-1 | |
| C8H4Cl2O4 | |
| 1 g | |
| ≥97.0% (T) | |
| 2,5-Dichloroterephthalic Acid |
| White-Yellow | |
| MFCD00058982 | |
| 235.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes