missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroterephthalic Acid 97.0+%, TCI America™

Catalog No. D16985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16985G 5 g
D16981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16985G Supplier TCI America Supplier No. D16985G

Chemical Identifiers

CAS 13799-90-1
Molecular Formula C8H4Cl2O4
MDL Number MFCD00058982

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 235.02
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloroterephthalic Acid