missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzylamine, 97%

Catalog No. p-7045782
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1197206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1197206 Supplier Thermo Scientific Chemicals Supplier No. L1197206

CAS: 10541-69-2 | C7H7Cl2N | 176.04 g/mol

2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Incompatible with strong oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 10541-69-2
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00052391
InChI Key AKGJLIXNRPNPCH-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzylamine, 2,5-dichlorophenyl methanamine, benzenemethanamine, 2,5-dichloro, 2,5-dichloro-benzylamine, 1-2,5-dichlorophenyl methanamine, 2,5-dichlorophenyl methylamine, 2,5-dichlorobenzylam, acmc-1bxya, rarechem al bw 0384, 2,5-dichlorobenzyl amine
PubChem CID 457600
IUPAC Name (2,5-dichlorophenyl)methanamine
SMILES C1=CC(=C(C=C1Cl)CN)Cl

Specifications

Density 1.318
Boiling Point 74°C to 76°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.5775
Quantity 5 g
UN Number UN2735
Beilstein 2639302
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only