Learn More
2,5-Dichlorobenzyl chloride, 97%
CAS: 2745-49-5 | C7H5Cl3 | 195.467 g/mol
$160.81 - $603.74
Chemical Identifiers
| CAS | 2745-49-5 |
|---|---|
| Molecular Formula | C7H5Cl3 |
| Molecular Weight (g/mol) | 195.467 |
| MDL Number | MFCD00035818 |
| InChI Key | OMZINLIPPVNUOG-UHFFFAOYSA-N |
| Synonym | 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component |
| PubChem CID | 75976 |
| IUPAC Name | 1,4-dichloro-2-(chloromethyl)benzene |
| SMILES | C1=CC(=C(C=C1Cl)CCl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAL0750606
|
Thermo Scientific Chemicals
L0750606 |
5 g |
Each for $160.81
|
|
|||||
|
AAL0750614
|
Thermo Scientific Chemicals
L0750614 |
25 g |
Each for $603.74
|
|
|||||
Description
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
Chemical Identifiers
| 2745-49-5 | |
| 195.467 | |
| OMZINLIPPVNUOG-UHFFFAOYSA-N | |
| 75976 | |
| C1=CC(=C(C=C1Cl)CCl)Cl |
| C7H5Cl3 | |
| MFCD00035818 | |
| 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component | |
| 1,4-dichloro-2-(chloromethyl)benzene |
Specifications
| 2745-49-5 | |
| Pungent | |
| 1.5755 | |
| 5 g | |
| 637802 | |
| Not miscible in water. | |
| C1=CC(=C(C=C1Cl)CCl)Cl | |
| 195.467 | |
| 195.48 | |
| 2,5-Dichlorobenzyl chloride |
| 121°C to 124°C (14 mmHg) | |
| C7H5Cl3 | |
| MFCD00035818 | |
| UN3265 | |
| 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component | |
| OMZINLIPPVNUOG-UHFFFAOYSA-N | |
| 1,4-dichloro-2-(chloromethyl)benzene | |
| 75976 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-382-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only