missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl chloride, 97%

Catalog No. AAL0750614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0750614 25 g
AAL0750606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0750614 Supplier Thermo Scientific Chemicals Supplier No. L0750614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2745-49-5 | C7H5Cl3 | 195.467 g/mol

2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.

Chemical Identifiers

CAS 2745-49-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00035818
InChI Key OMZINLIPPVNUOG-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component
PubChem CID 75976
IUPAC Name 1,4-dichloro-2-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)CCl)Cl

Specifications

Boiling Point 121°C to 124°C (14 mmHg)
Odor Pungent
Refractive Index 1.5755
Quantity 25 g
UN Number UN3265
Beilstein 637802
Solubility Information Not miscible in water.
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzyl chloride
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-382-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only