missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7127782
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35485G 5 g
D354825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35485G Supplier TCI America Supplier No. D35485G
Only null left

Chemical Identifiers

CAS 85482-13-9
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00236034
InChI Key UUVDOPTUDWJHFK-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
PubChem CID 3495744
IUPAC Name 2-(bromomethyl)-1,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)CBr)Cl

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 3261
Formula Weight 239.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorobenzyl Bromide