missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl alcohol, 99%

Catalog No. AAL0773406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0773406 5 g
AAL0773414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0773406 Supplier Thermo Scientific Chemicals Supplier No. L0773406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34145-05-6 | C7H6Cl2O | 177.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34145-05-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004607
InChI Key LCEIGNVIDJNUGF-UHFFFAOYSA-N
PubChem CID 118604
IUPAC Name (2,5-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)Cl

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Beilstein 3240770
Synonym 2,5-dichlorophenyl methanol, 2,5-dichlorobenzyl alcohol, benzenemethanol, 2,5-dichloro, 2,5-dichlorobenzylic alcohol, 2,5-dichloro-phenyl-methanol, 2,5-dichlorobenzylalcohol, acmc-1ad0a, 2,5-dichloro benzylalcohol, 2,5-dichloro-benzyl alcohol, benzenemethanol,2,5-dichloro
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,5-Dichlorobenzyl alcohol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 251-850-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only