missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7128053
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40185G 5 g
D401825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40185G Supplier TCI America Supplier No. D40185G
Only null left

Chemical Identifiers

CAS 21663-61-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00019741
InChI Key LNGWRTKJZCBXGT-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-dichloro, 2,5-dichloro-benzonitrile, 2,5-dichlorobenzonitrite, 2,5-dichlorobenzenecarbonitrile, pubchem3687, acmc-209fn5, ksc490q4h, 2,5-dichlorobenzonitrile, attercop-chm at133057
PubChem CID 89000
IUPAC Name 2,5-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)C#N)Cl

Specifications

Melting Point 131°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 172.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorobenzonitrile