missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126155
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D033825G 25 g
D0338100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D033825G Supplier TCI America Supplier No. D033825G
Only null left

Chemical Identifiers

CAS 50-79-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002416
InChI Key QVTQYSFCFOGITD-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dichloro, unii-kp7cmr0gsd, 2,5-dichlorobenzoicacid, kp7cmr0gsd, 2,5-dichloro-benzoic acid, ccris 8609, pubchem3685, 2,5dichlorobenzoic acid, wln: qvr bg eg, dsstox_cid_4978
PubChem CID 5784
IUPAC Name 2,5-dichlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorobenzoic Acid
EINECSNumber (3)-1410&(3)-1424
RTECSNumber DG6825000
TSCA Yes