missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzaldehyde 98.0+%, TCI America™

Catalog No. D28145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28145G Supplier TCI America Supplier No. D28145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6361-23-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00156140
InChI Key BUXHYMZMVMNDMG-UHFFFAOYSA-N
Synonym 2,5-dichloro-benzaldehyde, 2,5-dichlorobenzaldehdye, benzaldehyde, 2,5-dichloro, pubchem3081, 2.5-dichlorbenzaldehyd, 2,5-dichlorbenzaldehyde, acmc-1bkcw, 2,5-dichloro benzaldehdye, ksc493o7f, 2,5-dichlorobenzaldehyde
PubChem CID 80702
IUPAC Name 2,5-dichlorobenzaldehyde
SMILES ClC1=CC=C(Cl)C(C=O)=C1

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 233°C
Quantity 5 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorobenzaldehyde