missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzaldehyde, 98%

Catalog No. AAA1352506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1352506 5 g
AAA1352514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1352506 Supplier Thermo Scientific Chemicals Supplier No. A1352506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6361-23-5 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6361-23-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00156140
InChI Key BUXHYMZMVMNDMG-UHFFFAOYSA-N
Synonym 2,5-dichloro-benzaldehyde, 2,5-dichlorobenzaldehdye, benzaldehyde, 2,5-dichloro, pubchem3081, 2.5-dichlorbenzaldehyd, 2,5-dichlorbenzaldehyde, acmc-1bkcw, 2,5-dichloro benzaldehdye, ksc493o7f, 2,5-dichlorobenzaldehyde
PubChem CID 80702
IUPAC Name 2,5-dichlorobenzaldehyde
SMILES ClC1=CC=C(Cl)C(C=O)=C1

Specifications

Melting Point 53°C to 58°C
Boiling Point 231°C to 233°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN1759
Beilstein 1635476
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzaldehyde
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 228-832-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only