missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroanisole, TCI America™

Catalog No. D26055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26055G Supplier TCI America Supplier No. D26055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1984-58-3
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00061123
InChI Key QKMNFFSBZRGHDJ-UHFFFAOYSA-N
Synonym 2,5-dichloroanisole, benzene, 1,4-dichloro-2-methoxy, 2,5-dichloromethoxybenzene, banair, anisole, 2,5-dichloro, unii-8b3j0y3cj1, 2,5-dichloroanizole, pubchem3681, dsstox_cid_7452
PubChem CID 16125
IUPAC Name 1,4-dichloro-2-methoxybenzene
SMILES COC1=CC(Cl)=CC=C1Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 177.02
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dichloroanisole