missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroanisole, 98%

Catalog No. AAL1349706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1349706 5 g
AAL1349714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1349706 Supplier Thermo Scientific Chemicals Supplier No. L1349706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1984-58-3 | C7H6Cl2O | 177.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1984-58-3
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00061123
InChI Key QKMNFFSBZRGHDJ-UHFFFAOYSA-N
Synonym 2,5-dichloroanisole, benzene, 1,4-dichloro-2-methoxy, 2,5-dichloromethoxybenzene, banair, anisole, 2,5-dichloro, unii-8b3j0y3cj1, 2,5-dichloroanizole, pubchem3681, dsstox_cid_7452
PubChem CID 16125
IUPAC Name 1,4-dichloro-2-methoxybenzene
SMILES COC1=CC(Cl)=CC=C1Cl

Specifications

Density 1.33
Boiling Point 128°C to 132°C (3.5 mmHg)
Refractive Index 1.563
Quantity 5 g
Beilstein 2327401
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichloroanisole
Hazard Category H302+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-852-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only