missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-phenylenediamine, 99%

Catalog No. AAB2149406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2149406 5 g
AAB2149414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2149406 Supplier Thermo Scientific Chemicals Supplier No. B2149406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20103-09-7 | C6H6Cl2N2 | 177.028 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20103-09-7
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00007902
InChI Key QAYVHDDEMLNVMO-UHFFFAOYSA-N
Synonym 2,5-dichloro-1,4-phenylenediamine, 2,5-dichloro-p-phenylenediamine, 2,5-dichloro-1,4-benzenediamine, 1,4-diamino-2,5-dichlorobenzene, 1,4-benzenediamine, 2,5-dichloro, unii-9ccw77baxp, 1,4-diamino-3,6-dichlorobenzene, p-phenylenediamine, 2,5-dichloro, 9ccw77baxp, benzene, 2,5-dichloro, 1,4-diamino
PubChem CID 29949
IUPAC Name 2,5-dichlorobenzene-1,4-diamine
SMILES C1=C(C(=CC(=C1Cl)N)Cl)N

Specifications

Melting Point 164°C to 168°C
Quantity 5 g
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,5-Dichloro-p-phenylenediamine
Hazard Category H302
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 243-512-1
RTECSNumber SS9150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only