accessibility menu, dialog, popup

UserName

NH2ClNH2Cl

CAS RN 20103-09-7

IUPAC Name: 2,5-dichlorobenzene-1,4-diamine
Synonyms: 2,5-dichlorobenzene-1,4-diamine
Molecular Weight (g/mol): 177.03
Molecular Formula: C6H6Cl2N2
InChi Key: QAYVHDDEMLNVMO-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(N)C=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)