missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine 97.0+%, TCI America™
$136.08 - $402.63
Chemical Identifiers
| CAS | 761440-16-8 |
|---|---|
| Molecular Formula | C13H13Cl2N3O2S |
| Molecular Weight (g/mol) | 346.23 |
| MDL Number | MFCD12827956 |
| InChI Key | WWVLDJAVSQZSKO-UHFFFAOYSA-N |
| PubChem CID | 59764378 |
| IUPAC Name | 2,5-dichloro-N-[2-(propane-2-sulfonyl)phenyl]pyrimidin-4-amine |
| SMILES | CC(C)S(=O)(=O)C1=CC=CC=C1NC1=NC(Cl)=NC=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D5068200MG
|
TCI America
D5068200MG |
200 mg |
Each for $136.08
|
|
|||||
|
D50681G
|
TCI America
D50681G |
1 g |
Each for $402.63
|
|
|||||
Chemical Identifiers
| 761440-16-8 | |
| 346.23 | |
| WWVLDJAVSQZSKO-UHFFFAOYSA-N | |
| 2,5-dichloro-N-[2-(propane-2-sulfonyl)phenyl]pyrimidin-4-amine |
| C13H13Cl2N3O2S | |
| MFCD12827956 | |
| 59764378 | |
| CC(C)S(=O)(=O)C1=CC=CC=C1NC1=NC(Cl)=NC=C1Cl |
Specifications
| 761440-16-8 | |
| Yellow | |
| MFCD12827956 | |
| WWVLDJAVSQZSKO-UHFFFAOYSA-N | |
| 2,5-dichloro-N-[2-(propane-2-sulfonyl)phenyl]pyrimidin-4-amine | |
| 59764378 | |
| ≥97.0% (HPLC,N) | |
| 2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine |
| 151°C | |
| C13H13Cl2N3O2S | |
| 200 mg | |
| CC(C)S(=O)(=O)C1=CC=CC=C1NC1=NC(Cl)=NC=C1Cl | |
| 346.23 | |
| 346.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No