missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloro-4-nitroaniline 98.0+%, TCI America™
Supplier: TCI America D272225G
Specifications
| 2,5-Dichloro-4-nitroaniline | |
| 155°C | |
| C6H4Cl2N2O2 | |
| 25 g | |
| JBXZCPXEYAEMJS-UHFFFAOYSA-N | |
| 2,5-dichloro-4-nitroaniline | |
| 23108 | |
| ≥98.0% (GC) |
| 6627-34-5 | |
| Yellow | |
| MFCD00025157 | |
| benzenamine, 2,5-dichloro-4-nitro, 2,5-dichloro-4-nitro aniline, aniline, 2,5-dichloro-4-nitro, 1-amino-2,5-dichloro-4-nitrobenzene, 2,5-dichloro-4-nitrophenylamine, 2,5-dichloro-4-nitrobenzenamine, 2,5-dichloro-4-nitrobenzenenamine, acmc-1b7u0, 2,5-dichloro-p-nitroaniline, 2-12-00-00400 beilstein handbook reference | |
| C1=C(C(=CC(=C1Cl)[N+](=O)[O-])Cl)N | |
| 207.01 | |
| 207.01 | |
| Crystalline Powder |
Chemical Identifiers
| 6627-34-5 | |
| 207.01 | |
| JBXZCPXEYAEMJS-UHFFFAOYSA-N | |
| 23108 | |
| C1=C(C(=CC(=C1Cl)[N+](=O)[O-])Cl)N |
| C6H4Cl2N2O2 | |
| MFCD00025157 | |
| benzenamine, 2,5-dichloro-4-nitro, 2,5-dichloro-4-nitro aniline, aniline, 2,5-dichloro-4-nitro, 1-amino-2,5-dichloro-4-nitrobenzene, 2,5-dichloro-4-nitrophenylamine, 2,5-dichloro-4-nitrobenzenamine, 2,5-dichloro-4-nitrobenzenenamine, acmc-1b7u0, 2,5-dichloro-p-nitroaniline, 2-12-00-00400 beilstein handbook reference | |
| 2,5-dichloro-4-nitroaniline |
Safety and Handling
EINECSNumber : (3)-0423
RTECSNumber : BX2950000
TSCA : Yes