missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-4-nitroaniline 98.0+%, TCI America™

Catalog No. D27225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27225G 5 g
D272225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27225G Supplier TCI America Supplier No. D27225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6627-34-5
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00025157
InChI Key JBXZCPXEYAEMJS-UHFFFAOYSA-N
Synonym benzenamine, 2,5-dichloro-4-nitro, 2,5-dichloro-4-nitro aniline, aniline, 2,5-dichloro-4-nitro, 1-amino-2,5-dichloro-4-nitrobenzene, 2,5-dichloro-4-nitrophenylamine, 2,5-dichloro-4-nitrobenzenamine, 2,5-dichloro-4-nitrobenzenenamine, acmc-1b7u0, 2,5-dichloro-p-nitroaniline, 2-12-00-00400 beilstein handbook reference
PubChem CID 23108
IUPAC Name 2,5-dichloro-4-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)[N+](=O)[O-])Cl)N

Specifications

Melting Point 155°C
Color Yellow
Quantity 5 g
Formula Weight 207.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-4-nitroaniline
EINECSNumber (3)-0423
RTECSNumber BX2950000
TSCA Yes