missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloro-3-methylthiophene 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 185°C |
| Quantity | 5 g |
| Formula Weight | 167.05 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,5-Dichloro-3-methylthiophene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |