accessibility menu, dialog, popup

UserName

CH3ClSCl

CAS RN 17249-90-0

IUPAC Name: 2,5-dichloro-3-methylthiophene
Synonyms: 2,5-dichloro-3-methylthiophene
Molecular Weight (g/mol): 167.05
Molecular Formula: C5H4Cl2S
InChi Key: BUWSFDLUTRHPBO-UHFFFAOYSA-N
SMILES: CC1=C(Cl)SC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results