missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-3-methylthiophene 96.0+%, TCI America™

Catalog No. D36801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36801G 1 g
D36805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36801G Supplier TCI America Supplier No. D36801G
Only null left

Chemical Identifiers

CAS 17249-90-0
Molecular Formula C5H4Cl2S
Molecular Weight (g/mol) 167.05
MDL Number MFCD00022490
InChI Key BUWSFDLUTRHPBO-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-methyl thiophene, 2,5-dichloro-3-methyl-thiophene, 2,5-dichlor-3-methylthiophen, pubchem5195, acmc-209e4q, 2,5-dichloro-3-methylthiophene
PubChem CID 268910
IUPAC Name 2,5-dichloro-3-methylthiophene
SMILES CC1=C(Cl)SC(Cl)=C1

Specifications

Color Yellow
Boiling Point 185°C
Quantity 1 g
Formula Weight 167.05
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dichloro-3-methylthiophene
TSCA No
Recommended Storage Refrigerator