missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloro-1,4-phenylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 166°C |
| Quantity | 25 g |
| Formula Weight | 177.03 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Dichloro-1,4-phenylenediamine |
Safety and Handling
| EINECSNumber | (3)-0295 |
| RTECSNumber | SS9150000 |
| TSCA | Yes |