missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals

Catalog No. AC263480250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263480250 25 g Glass bottle
AC263480050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC263480250 Supplier Thermo Scientific Chemicals Supplier No. 263480250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 615-59-8 | Molecular Formula: C7H6Br2 | Molecular Weight: 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-59-8
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000090
InChI Key QKEZTJYRBHOKHH-UHFFFAOYSA-N
Synonym 2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
PubChem CID 12006
IUPAC Name 1,4-dibromo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Colorless to Yellow
Density 1.8100g/mL
Boiling Point 135.0°C to 136.0°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6010 to 1.6030
Linear Formula CH3C6H3Br2
Quantity 25 g
Beilstein 05, 308
Specific Gravity 1.81
Solubility Information Solubility in water: immiscible
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Dibromotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 210-437-0
TSCA TSCA

RUO – Research Use Only