missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromotoluene, 98%

Catalog No. p-7024555
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1941806 5 g
AAA1941814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1941806 Supplier Thermo Scientific Chemicals Supplier No. A1941806
Only null left

CAS: 615-59-8 | C7H6Br2 | 249.933 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-59-8
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000090
InChI Key QKEZTJYRBHOKHH-UHFFFAOYSA-N
Synonym 2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
PubChem CID 12006
IUPAC Name 1,4-dibromo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Br

Specifications

Melting Point 5°C to 6°C
Density 1.813
Boiling Point 106°C to 108°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Aromatic
Refractive Index 1.602
Quantity 5 g
Beilstein 1859123
Formula Weight 249.94
Percent Purity 98%
Chemical Name or Material 2,5-Dibromotoluene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-437-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only