missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothieno[3,2-b]thiophene 98.0+%, TCI America™

Catalog No. p-7127858
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3678200MG 200 mg
D36781G 1 g
D36785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D3678200MG Supplier TCI America Supplier No. D3678200MG
Only null left

Chemical Identifiers

CAS 25121-87-3
Molecular Formula C6H2Br2S2
Molecular Weight (g/mol) 298.01
MDL Number MFCD03931293
InChI Key APDAUBNBDJUQGW-UHFFFAOYSA-N
PubChem CID 3597455
IUPAC Name 2,5-dibromothieno[3,2-b]thiophene
SMILES C1=C(SC2=C1SC(=C2)Br)Br

Specifications

Melting Point 130°C
Quantity 200 mg
Formula Weight 298.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dibromothieno[3,2-b]thiophene
TSCA No
Recommended Storage Freezer