accessibility menu, dialog, popup

UserName

CAS RN 25121-87-3

IUPAC Name: 2,5-dibromothieno[3,2-b]thiophene
Synonyms: 2,5-dibromothieno[3,2-b]thiophene
Molecular Weight (g/mol): 298.01
Molecular Formula: C6H2Br2S2
InChi Key: APDAUBNBDJUQGW-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(S1)C=C(Br)S2
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results